5-Nitro-α,α,α-trifluoro-m-toluic acid

Supplier: TCI America

Synonyms: 3-Nitro-5-(trifluoromethyl)benzoic acid

T1344-25G T1344-5G
TCT1344-25GEA 458 USD
TCT1344-25G TCT1344-005G
5-Nitro-α,α,α-trifluoro-m-toluic acid
5-Nitro-α,α,α-trifluoro-m-toluic acid

CAS Number: 328-80-3
MDL Number: MFCD00024509
Molecular Formula: C8H4F3NO4
Molecular Weight: 235.12
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 129


Other Product Names / Synonyms: 3-Carboxy-5-nitrobenzotrifluoride

Formula: C₈H₄F₃NO₄
MW: 235.12 g/mol
Melting Pt: 129 °C
MDL Number: MFCD00024509
CAS Number: 328-80-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2432091700eca3df39dab96246a6396bf80b315f&vid=2432091700eca3df39dab96246a6396bf80b315f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR