Tetrabutylammonium fluoride ∼1 mol/L in THF

Supplier: TCI America

T1338-500ML T1338-100ML T1338-25ML
TCT1338-500MLEA 192.5 USD
TCT1338-500ML TCT1338-100ML TCT1338-25ML
Tetrabutylammonium fluoride ∼1 mol/L in THF
Tetrabutylammonium fluoride

CAS Number: 429-41-4
MDL Number: MFCD00011747
Molecular Formula: C16H36FN
Molecular Weight: 261.47
Form: Clear Liquid
Color: Colorless
Flash Point (°C): -17
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C


Other Product Names / Synonyms: TBAF

Formula: C₁₆H₃₆FN
MW: 261.47 g/mol
Density: 0.92 (20 °C)
Flash Pt: -17 °C
MDL Number: MFCD00011747
CAS Number: 429-41-4
EINECS: 207-057-2
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e372eed26b80831c6e6740c7e3ea12856bd7382&vid=1eb0eb42c4d48aa9bdcb1e2ebcb684fda2d180fc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR