Methyl mercaptoacetate ≥98.0%

Supplier: TCI America

Synonyms: Methyl thioglycolate

T1287-500G T1287-25G
TCT1287-500GEA 149 USD
TCT1287-500G TCT1287-025G
Methyl mercaptoacetate ≥98.0%
Methyl mercaptoacetate

CAS Number: 2365-48-2
MDL Number: MFCD00004873
Molecular Formula: C3H6O2S
Molecular Weight: 106.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 49
Flash Point (°C): 51
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: Mercaptoacetic Acid Methyl Ester, Methyl Mercaptoacetate, Thioglycolic Acid Methyl Ester

Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 49 °C
Density: 1.17 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00004873
CAS Number: 2365-48-2
EINECS: 219-121-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcb2558fd8203305953102c31b1f6cd95f1a2a92&vid=bcb2558fd8203305953102c31b1f6cd95f1a2a92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR