Tetrabutylammonium tribromide ≥98.0%

Supplier: TCI America
Warning

Synonyms: Tetra-n-butylammonium tribromide, N,N,N,N-Tetrabutylammonium tribromide

T1284-500G T1284-25G T1284-100G
TCT1284-500GEA 289 USD
TCT1284-500G TCT1284-25G TCT1284-100G
Tetrabutylammonium tribromide ≥98.0%
Tetrabutylammonium tribromide

CAS Number: 38932-80-8
MDL Number: MFCD00012110
Molecular Formula: C16H36Br3N
Molecular Weight: 482.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Yellow
Melting point (°C): 73

Formula: C₁₆H₃₆Br₃N
MW: 482.18 g/mol
Melting Pt: 76
MDL Number: MFCD00012110
CAS Number: 38932-80-8
UN: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c20785fa74f0c70d7140507c43a1c104da3ab7a&vid=0c20785fa74f0c70d7140507c43a1c104da3ab7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR