2,3,4-Trichloronitrobenzene ≥99.0%
Supplier: TCI America
Synonyms:
1-Nitro-2,3,4-trichlorobenzene, 1,2,3-Trichloro-4-nitrobenzene, 4-Nitro-1,2,3-trichlorobenzene
T1197-500G
T1197-25G
TCT1197-500GEA
408
USD
TCT1197-500G
TCT1197-025G
2,3,4-Trichloronitrobenzene ≥99.0%
2,3,4-Trichloronitrobenzene
CAS Number: 17700-09-3
MDL Number: MFCD00007063
Molecular Formula: C6H2Cl3NO2
Molecular Weight: 226.44
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 56
Flash Point (°C): 110
Formula:
C₆H₂Cl₃NO₂ MW: 226.45 g/mol Melting Pt: 56 °C Flash Pt: 110 °C |
MDL Number:
MFCD00007063 CAS Number: 17700-09-3 EINECS: 241-705-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...