D-(+)-Tyrosine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-D-Tyr-OH

T1141-5G T1141-25G
TCT1141-005GEA 51.58 USD
TCT1141-005G TCT1141-025G
D-(+)-Tyrosine ≥98.0% (by titrimetric analysis)
D-(+)-Tyrosine

CAS Number: 556-02-5
MDL Number: MFCD00063073
Molecular Formula: C9H11NO3
Molecular Weight: 181.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 314
Specific rotation [a]20/D: 11.2 deg (C=5, 1mol/L HCl)


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Tyr-OH

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 314 °C
MDL Number: MFCD00063073
CAS Number: 556-02-5
EINECS: 209-112-6
Merck Index: 12,09970
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bf020525ab674e977ad157aeb90f69f1ede08ea&vid=8bf020525ab674e977ad157aeb90f69f1ede08ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR