Tetrabutoxysilane ≥98.0%

Supplier: TCI America
Warning

Synonyms: Tetrabutyl orthosilicate

T1126-500ML T1126-25ML
TCT1126-500MLEA 307.3 USD
TCT1126-500ML TCT1126-25ML
Tetrabutoxysilane ≥98.0%
Tetrabutoxysilane

CAS Number: 4766-57-8
MDL Number: MFCD00009432
Molecular Formula: C16H36O4Si
Molecular Weight: 320.55
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 277
Flash Point (°C): 136
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: Tetrabutoxysilane

Formula: C₁₆H₃₆O₄Si
MW: 320.54 g/mol
Boiling Pt: 277 °C
Density: 0.90 (20 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00009432
CAS Number: 4766-57-8
EINECS: 225-305-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=172d03f06940789cb5228ffcdecd231cf73e7a29&vid=172d03f06940789cb5228ffcdecd231cf73e7a29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR