Tetrabutylphosphonium bromide ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetra-n-butylphosphonium bromide

T1124-500G T1124-25G T1124-100G
TCT1124-500GEA 298 USD
TCT1124-500G TCT1124-25G TCT1124-100G
Tetrabutylphosphonium bromide ≥99.0% (by titrimetric analysis)
Tetrabutylphosphonium bromide

CAS Number: 3115-68-2
MDL Number: MFCD00011853
Molecular Formula: C16H36BrP
Molecular Weight: 339.34
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Melting point (°C): 104
Flash Point (°C): 290

Formula: C₁₆H₃₆BrP
MW: 339.34 g/mol
Melting Pt: 104 °C
Flash Pt: 290 °C
MDL Number: MFCD00011853
CAS Number: 3115-68-2
EINECS: 221-487-8
UN: 3464
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6929090fdcfed1067fdc5cbc8c9ba3d9a7a2fb7&vid=d6929090fdcfed1067fdc5cbc8c9ba3d9a7a2fb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR