2',4',6'-Trimethylacetophenone ≥97.0%
Supplier: TCI America
Synonyms:
1-(2,4,6-Trimethylphenyl)ethan-1-one
T1117-5ML
T1117-25ML
TCT1117-005MLEA
30.53
USD
TCT1117-005ML
TCT1117-025ML
2',4',6'-Trimethylacetophenone ≥97.0%
2',4',6'-Trimethylacetophenone
CAS Number: 1667-01-2
MDL Number: MFCD00008735
Molecular Formula: C11H14O
Molecular Weight: 162.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 236
Flash Point (°C): 110
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 2-Acetylmesitylene
Formula:
C₁₁H₁₄O MW: 162.23 g/mol Boiling Pt: 236 °C Density: 0.98 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00008735 CAS Number: 1667-01-2 EINECS: 216-783-9 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...