2,2,6-Trimethyl-4H-1,3-dioxin-4-one ≥95.0% (by NMR)

Supplier: TCI America

T1099-500ML T1099-25ML
TCT1099-500MLEA 194.25 USD
TCT1099-500ML TCT1099-025ML
2,2,6-Trimethyl-4H-1,3-dioxin-4-one ≥95.0% (by NMR)
2,2,6-Trimethyl-4H-1,3-dioxin-4-one

CAS Number: 5394-63-8
MDL Number: MFCD00040468
Molecular Formula: C7H10O3
Molecular Weight: 142.15
Purity/Analysis Method: >95.0% (NMR)
Form: Liquid
Color: Yellow
Boiling point (°C): 77
Flash Point (°C): 86
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: 2,2,4-Trimethyl-6-keto-1,3-dioxine, Diketene Acetone Adduct

Formula: C₇H₁₀O₃
Boiling Pt: 77 °C
Density: 1.08 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00040468
CAS Number: 5394-63-8
EINECS: 226-403-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf5e90595169b6d16a88bd5a54be27eda89835c1&vid=bf5e90595169b6d16a88bd5a54be27eda89835c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR