Triisopropylchlorosilane ≥95.0%

Supplier: TCI America
Danger

Synonyms: Chlorotriisopropylsilane, Triisopropylsilyl chloride, Triisopropylchlorosilane

T1078-25ML T1078-5ML T1078-250ML
TCT1078-025MLEA 118.29 USD
TCT1078-025ML TCT1078-005ML TCT1078-250ML
Triisopropylchlorosilane ≥95.0%
Triisopropylchlorosilane

CAS Number: 13154-24-0
MDL Number: MFCD00009656
Molecular Formula: C9H21ClSi
Molecular Weight: 192.80
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 96
Flash Point (°C): 62
Specific Gravity (20/20): 0.91


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis), Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Chlorotriisopropylsilane

Formula: C₉H₂₁ClSi
MW: 192.8 g/mol
Boiling Pt: 96 °C
Density: 0.91 (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00009656
CAS Number: 13154-24-0
UN: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d03420a3a6b000c0a33940d8fec422b04036ec4&vid=5d03420a3a6b000c0a33940d8fec422b04036ec4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR