Tetraphenylphosphonium bromide ≥98.0%

Supplier: TCI America

T1069-500G T1069-25G
TCT1069-500GEA 175 USD
TCT1069-500G TCT1069-025G
Tetraphenylphosphonium bromide ≥98.0%
Tetraphenylphosphonium bromide

CAS Number: 2751-90-8
MDL Number: MFCD00011915
Molecular Formula: C24H20BrP
Molecular Weight: 419.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₂₄H₂₀BrP
MW: 419.3 g/mol
Melting Pt: 299…300 °C
MDL Number: MFCD00011915
CAS Number: 2751-90-8
EINECS: 220-393-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=927445765c2aca17bbb89e9dba6b3bdf13da07e4&vid=927445765c2aca17bbb89e9dba6b3bdf13da07e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR