3,4,5-Trimethoxyaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
T1055-5G
T1055-25G
TCT1055-005GEA
69
USD
TCT1055-005G
TCT1055-025G
3,4,5-Trimethoxyaniline ≥98.0% (by GC, titration analysis)
3,4,5-Trimethoxyaniline
CAS Number: 24313-88-0
MDL Number: MFCD00008393
Molecular Formula: C9H13NO3
Molecular Weight: 183.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 112
Formula:
C₉H₁₃NO₃ MW: 183.21 g/mol Boiling Pt: 293 °C (1013 hPa) Melting Pt: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008393 CAS Number: 24313-88-0 EINECS: 246-154-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...