Tetrapropylammonium hydroxide 20 - 25% in water

Supplier: TCI America
Danger

T1020-500ML T1020-25ML T1020-100ML
TCT1020-500MLEA 156.35 USD
TCT1020-500ML TCT1020-25ML TCT1020-100ML
Tetrapropylammonium hydroxide 20 - 25% in water
Tetrapropylammonium hydroxide

CAS Number: 4499-86-9
MDL Number: MFCD00009360
Molecular Formula: C12H29NO
Molecular Weight: 203.37
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.01

Formula: C₁₂H₂₉NO
Density: 1.01 (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009360
CAS Number: 4499-86-9
EINECS: 224-800-6
UN: 3267
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=453b68e0acc5ce0e39edc269272904ffe0944ec6&vid=31d9e72df41ea6203fb817e211de8f94161d6708&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Concentration (Neutralization titration) 20.0 to 25.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR