trans-2-Methyl-2-butenal ≥95.0%

Supplier: TCI America
Danger

Synonyms: Trans-2-methyl-2-butenal, Tiglic aldehyde, (E)-2-Methyl-2-butenal

T1003-5ML T1003-25ML
TCT1003-005MLEA 34.08 USD
TCT1003-005ML TCT1003-025ML
trans-2-Methyl-2-butenal ≥95.0%
trans-2-Methyl-2-butenal

CAS Number: 497-03-0
MDL Number: MFCD00006977
Molecular Formula: C5H8O
Molecular Weight: 84.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 116
Flash Point (°C): 18
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Hemiterpenes
Other Product Names / Synonyms: Tiglic Aldehyde

Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 116 °C
Density: 0.87 (20 °C)
Flash Pt: 18 °C
MDL Number: MFCD00006977
CAS Number: 497-03-0
EINECS: 207-833-0
UN: 1989
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba99409d6c96c5761fb9ba051dc19ea2f6e16b49&vid=8eff4c299086ec975e0c1a6d9bb02e5dfbf75e11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR