Tetrapropylammonium hydroxide 10% in water
Supplier: TCI America
|
Danger
|
T0964-500ML
T0964-25ML
T0964-100ML
TCT0964-500MLEA
114
USD
TCT0964-500ML
TCT0964-25ML
TCT0964-100ML
Tetrapropylammonium hydroxide 10% in water
Tetrapropylammonium hydroxide
CAS Number: 4499-86-9
MDL Number: MFCD00009360
Molecular Formula: C12H29NO
Molecular Weight: 203.37
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.00
Formula:
C₁₂H₂₉NO Density: 1.00 (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009360 CAS Number: 4499-86-9 EINECS: 224-800-6 UN: 3267 ADR: 8,III |
|
Specification Test Results
Appearance | Colourless to Almost Colourless clear liquid |
Concentration (Neutralization titration) | 10.0 to 11.0 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...