Trimethyltin chloride ≥98.0%

Supplier: TCI America
Danger

T0958-25G T0958-5G
TCT0958-025GEA 231 USD
TCT0958-025G TCT0958-005G
Trimethyltin chloride ≥98.0%
Trimethyltin chloride

CAS Number: 1066-45-1
MDL Number: MFCD00000520
Molecular Formula: C3H9ClSn
Molecular Weight: 199.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 156
Melting point (°C): 40
Flash Point (°C): 97
Storage Temperature: 0-10°C

Formula: C₃H₉ClSn
Boiling Pt: 156 °C
Melting Pt: 40 °C
Flash Pt: 97 °C
MDL Number: MFCD00000520
CAS Number: 1066-45-1
EINECS: 213-917-8
UN: 3146
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab75f552a4ba450b662c1652bf3d6d210b5cd50e&vid=ab75f552a4ba450b662c1652bf3d6d210b5cd50e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR