1,3,3-Trimethyl-2-methyleneindoline ≥96.0%
Supplier: TCI America
CAS Number: 118-12-7
MDL Number: MFCD00005813
Molecular Formula: C12H15N
Molecular Weight: 173.26
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Red
Boiling point (°C): 248
Melting point (°C): -10
Flash Point (°C): 101
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: Fischer's Base
Formula:
C₁₂H₁₅N Boiling Pt: 248 °C Melting Pt: –10 °C Density: 0.98 (20 °C) Flash Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005813 CAS Number: 118-12-7 EINECS: 204-235-1 UN: 3082 ADR: 9,III |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...