Tetradecylbenzene ≥97.0%
Supplier: TCI America
CAS Number: 1459-10-5
MDL Number: MFCD00015080
Molecular Formula: C20H34
Molecular Weight: 274.49
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 207
Melting point (°C): 16
Flash Point (°C): 112
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: Myristylbenzene, 1-Phenyltetradecane
Formula:
C₂₀H₃₄ Boiling Pt: 207 °C Melting Pt: 16 °C Density: 0.86 (20 °C) Flash Pt: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015080 CAS Number: 1459-10-5 EINECS: 215-950-3 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...