Tetrabutylammonium perchlorate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetra-n-butylammonium perchlorate

T0836-500G T0836-25G
TCT0836-500GEA 511.6 USD
TCT0836-500G TCT0836-25G
Tetrabutylammonium perchlorate ≥98.0% (by titrimetric analysis)
Tetrabutylammonium perchlorate

CAS Number: 1923-70-2
MDL Number: MFCD00038722
Molecular Formula: C16H36ClNO4
Molecular Weight: 341.92
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₁₆H₃₆ClNO₄
MW: 341.92 g/mol
Melting Pt: 210 °C
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00038722
CAS Number: 1923-70-2
EINECS: 217-655-5
UN: 1481
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=297a6a3b04a7287f82b5518dce8db8a79437fe51&vid=297a6a3b04a7287f82b5518dce8db8a79437fe51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR