Terephthalic acid dihydrazide ≥90.0% (by HPLC)
Supplier: TCI America
Synonyms:
Terephthalic Dihydrazide
T0758-25G
T0758-500G
TCT0758-025GEA
97
USD
TCT0758-025G
TCT0758-500G
Terephthalic acid dihydrazide ≥90.0% (by HPLC)
Terephthalic acid dihydrazide
CAS Number: 136-64-1
MDL Number: MFCD00014762
Molecular Formula: C8H10N4O2
Molecular Weight: 194.19
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Color: White
Other Product Names / Synonyms: Terephthalohydrazide
Formula:
C₈H₁₀N₄O₂ MW: 194.19 g/mol |
MDL Number:
MFCD00014762 CAS Number: 136-64-1 EINECS: 205-253-2 |
Specification Test Results
Purity/Analysis Method | >90.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...