3,4,5-Trimethoxybenzamide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
T0718-10G
T0718-25G
TCT0718-010GEA
52
USD
TCT0718-010G
TCT0718-25G
3,4,5-Trimethoxybenzamide ≥98.0% (by HPLC, total nitrogen)
3,4,5-Trimethoxybenzamide
CAS Number: 3086-62-2
MDL Number: MFCD00017129
Molecular Formula: C10H13NO4
Molecular Weight: 211.22
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 180
Formula:
C₁₀H₁₃NO₄ MW: 211.22 g/mol Melting Pt: 180 °C |
MDL Number:
MFCD00017129 CAS Number: 3086-62-2 EINECS: 221-406-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...