Tropinone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 8-Methyl-8-azabicyclo[3.2.1]octan-3-one

T0626-25G T0626-5G
TCT0626-25GEA 123 USD
TCT0626-25G TCT0626-005G
Tropinone ≥98.0% (by GC, titration analysis)
Tropinone

CAS Number: 532-24-1
MDL Number: MFCD00005549
Molecular Formula: C8H13NO
Molecular Weight: 139.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 218
Melting point (°C): 43
Flash Point (°C): 90


Other Product Names / Synonyms: 3-Tropanone

Formula: C₈H₁₃NO
MW: 139.2 g/mol
Boiling Pt: 218 °C
Melting Pt: 43 °C
Flash Pt: 90 °C
MDL Number: MFCD00005549
CAS Number: 532-24-1
EINECS: 208-530-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a326c14796d4cfe8f69f23119b80f114b9b969c3&vid=a326c14796d4cfe8f69f23119b80f114b9b969c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR