2,2,2-Trichloroethanol ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
2,2,2-Trichloroethan-1-ol
T0622-500G
T0622-25G
TCT0622-500GEA
148
USD
TCT0622-500G
TCT0622-25G
2,2,2-Trichloroethanol ≥98.0%
2,2,2-Trichloroethanol
CAS Number: 115-20-8
MDL Number: MFCD00004677
Molecular Formula: C2H3Cl3O
Molecular Weight: 149.40
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 151
Melting point (°C): 18
Flash Point (°C): 110
Specific Gravity (20/20): 1.56
Formula:
C₂H₃Cl₃O MW: 149.4 g/mol Boiling Pt: 151 °C Melting Pt: 18 °C Density: 1.56 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004677 CAS Number: 115-20-8 EINECS: 204-071-0 UN: 2810 ADR: 6.1,III Merck Index: 12,09786 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...