Tetraethylurea ≥99.0%

Supplier: TCI America

T0587-25ML T0587-100ML
TCT0587-025MLEA 87 USD
TCT0587-025ML 76220-488
Tetraethylurea ≥99.0%
Tetraethylurea

CAS Number: 1187-03-7
MDL Number: MFCD00042881
Molecular Formula: C9H20N2O
Molecular Weight: 172.27
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 214
Flash Point (°C): 81
Specific Gravity (20/20): 0.91

Formula: C₉H₂₀N₂O
Boiling Pt: 214 °C
Density: 0.91 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00042881
CAS Number: 1187-03-7
EINECS: 214-696-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7d71f06cd0c9c715404b8ea79ccf51a4c0c9d66&vid=b7d71f06cd0c9c715404b8ea79ccf51a4c0c9d66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR