1-(Trimethylsilyl)imidazole ≥98.0%

Supplier: TCI America

Synonyms: TSIM, N-(Trimethylsilyl)imidazole, N-(Trimethylsilyl)-1H-imidazole, TMSI, 1-(Trimethylsilyl)-1H-imidazole

T0585-100G T0585-25G
TCT0585-100GEA 130.47 USD
TCT0585-100G TCT0585-25G
1-(Trimethylsilyl)imidazole ≥98.0%
1-(Trimethylsilyl)imidazole

[Trimethylsilylating Reagent]
CAS Number: 18156-74-6
MDL Number: MFCD00005280
Molecular Formula: C6H12N2Si
Molecular Weight: 140.26
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 91
Flash Point (°C): 5
Specific Gravity (20/20): 0.96
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis), Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: TMS-Imidazole, SIM, TMSI

Formula: C₆H₁₂N₂Si
Boiling Pt: 91 °C
Melting Pt: –42 °C
Density: 0.96 (20 °C)
Flash Pt: 5 °C
MDL Number: MFCD00005280
CAS Number: 18156-74-6
EINECS: 242-040-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a343d14a9972e232921a6ab4b799fbc582c5cf95&vid=4135019109ea889cc6b1ac753f3ad090c9fa641b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR