DL-Tyrosine ≥98.0%

Supplier: TCI America

Synonyms: 4-Hydroxyphenylalanine, 3-(p-Hidroxifenil)-DL-alanina, H-DL-Tyr-OH

T0549-25G T0549-5G
TCT0549-025GEA 84 USD
TCT0549-025G TCT0549-005G
DL-Tyrosine ≥98.0%
DL-Tyrosine

CAS Number: 556-03-6
MDL Number: MFCD00063074
Molecular Formula: C9H11NO3
Molecular Weight: 181.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Tyr-OH

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 378
Melting Pt: 285
MDL Number: MFCD00063074
CAS Number: 556-03-6
EINECS: 209-113-1
Merck Index: 12,09970
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3508d37abec67ab7dea4ee8aa68c1b6a75b7891&vid=e3508d37abec67ab7dea4ee8aa68c1b6a75b7891&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR