1,6-Bis(dimethylamino)hexane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T0537-25ML T0537-500ML
TCT0537-25MLEA 34.58 USD
TCT0537-25ML TCT0537-500ML
1,6-Bis(dimethylamino)hexane ≥98.0% (by GC, titration analysis)
1,6-Bis(dimethylamino)hexane

CAS Number: 111-18-2
MDL Number: MFCD00008339
Molecular Formula: C10H24N2
Molecular Weight: 172.32
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 210
Flash Point (°C): 75
Specific Gravity (20/20): 0.80


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: 1,6-Bis(dimethylamino)hexane, N,N,N',N'-Tetramethyl-1,6-hexanediamine

Formula: C₁₀H₂₄N₂
Boiling Pt: 210 °C
Melting Pt: -46 °C
Density: 0.80 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00008339
CAS Number: 111-18-2
EINECS: 203-842-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e2b578d0ac95c9e5ac94fb142f34244df6c0c81&vid=5e2b578d0ac95c9e5ac94fb142f34244df6c0c81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR