Tropine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol
T0534-5G
T0534-25G
TCT0534-005GEA
32.56
USD
TCT0534-005G
TCT0534-025G
Tropine ≥98.0% (by GC, titration analysis)
Tropine
CAS Number: 120-29-6
MDL Number: MFCD00005551
Molecular Formula: C8H15NO
Molecular Weight: 141.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Product Categories: Bioscience, Tropane Alkaloids
Other Product Names / Synonyms: Tropanol
Formula:
C₈H₁₅NO MW: 141.21 g/mol |
MDL Number:
MFCD00180667 CAS Number: 120-29-6 EINECS: 204-384-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...