2,3,5-Triphenyltetrazolium chloride (TTC) ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC
T0520-25G
T0520-1G
TCT0520-025GEA
223.91
USD
TCT0520-025G
TCT0520-001G
2,3,5-Triphenyltetrazolium chloride (TTC) ≥98.0%
2,3,5-Triphenyltetrazolium chloride (TTC)
CAS Number: 298-96-4
MDL Number: MFCD00011963
Molecular Formula: C19H15ClN4
Molecular Weight: 334.81
Purity/Analysis Method: >98.0% (T)
Form: Powder
Color: White
Other Product Names / Synonyms: Tetrazole Red, Tetrazolium Red, TTC
Formula:
C₁₉H₁₅ClN₄ MW: 334.81 g/mol Boiling Pt: ∼360 °C (1013 hPa) Melting Pt: 235 Storage Temperature: Refrigerator |
MDL Number:
MFCD00011963 CAS Number: 298-96-4 EINECS: 206-071-6 Merck Index: 13,09815 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...