Triphenylmethane ≥98.0%

Supplier: TCI America

T0515-500G T0515-25G T0515-100G
TCT0515-500GEA 365 USD
TCT0515-500G TCT0515-025G TCT0515-100G
Triphenylmethane ≥98.0%
Triphenylmethane

CAS Number: 519-73-3
MDL Number: MFCD00004763
Molecular Formula: C19H16
Molecular Weight: 244.34
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 359
Melting point (°C): 94
Flash Point (°C): 100

Formula: C₁₉H₁₆
MW: 244.34 g/mol
Boiling Pt: 359 °C
Melting Pt: 94 °C
Density: 1.01 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004763
CAS Number: 519-73-3
EINECS: 208-275-0
Merck Index: 12,09871
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fdc269a20dbe1c0ed26474abf432d562ab4493b&vid=2fdc269a20dbe1c0ed26474abf432d562ab4493b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR