Triphenyl phosphite ≥97.0%

Supplier: TCI America
Warning

T0510-500G T0510-25G
TCT0510-500GEA 37.62 USD
TCT0510-500G TCT0510-25G
Triphenyl phosphite ≥97.0%
Triphenyl phosphite

CAS Number: 101-02-0
MDL Number: MFCD00003032
Molecular Formula: C18H15O3P
Molecular Weight: 310.29
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 360
Flash Point (°C): 206
Freezing point (°C): 20
Specific Gravity (20/20): 1.19


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Phosphorous Acid Triphenyl Ester

Formula: C₁₈H₁₅O₃P
MW: 310.29 g/mol
Boiling Pt: 360 °C
Melting Pt: 22…25 °C
Density: 1.19 (20 °C)
Flash Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00003032
CAS Number: 101-02-0
EINECS: 202-908-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e542cd734cf793f64c49397758c281520d7b5321&vid=e542cd734cf793f64c49397758c281520d7b5321&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR