Trimethylamine ∼28% ∼4.3 mol/L in water

Supplier: TCI America
Danger

T0464-25ML T0464-500ML
TCT0464-25MLEA 24.62 USD
TCT0464-25ML TCT0464-500ML
Trimethylamine ∼28% ∼4.3 mol/L in water
Trimethylamine

CAS Number: 75-50-3
MDL Number: MFCD00008327
Molecular Formula: C3H9N
Molecular Weight: 59.11
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 3
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C

Formula: C₃H₉N
MW: 59.11 g/mol
Boiling Pt: 30 °C (1013 hPa)
Melting Pt: -3 °C
Density: 0.92 (20 °C)
Flash Pt: 3 °C
Storage Temperature: Ambient
MDL Number: MFCD00008327
CAS Number: 75-50-3
EINECS: 200-875-0
UN: 1297
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5591e14b9e32e2d34918ad382bcd7050f23cb5f&vid=f5b8f4a12cb7334a45c577c9c38879a250788549&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR