1-(2-Thenoyl)-3,3,3-trifluoroacetone ≥98.0%
Supplier: TCI America
Synonyms:
(TTA), Thenoyltrifluoroacetone, 4,4,4-Trifluoro-1-(2-thienyl)butane-1,3-dione, TTFA, 2-Thenoyltrifluoroacetone, (2-Thenoyl)-ω,ω,ω-trifluoroacetone
T0438-500G
T0438-25G
TCT0438-500GEA
399
USD
TCT0438-500G
TCT0438-025G
1-(2-Thenoyl)-3,3,3-trifluoroacetone ≥98.0%
1-(2-Thenoyl)-3,3,3-trifluoroacetone
CAS Number: 326-91-0
MDL Number: MFCD00005445
Molecular Formula: C8H5F3O2S
Molecular Weight: 222.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 104
Melting point (°C): 45
Flash Point (°C): 111
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 2-Thenoyltrifluoroacetone
Formula:
C₈H₅F₃O₂S MW: 222.19 g/mol Boiling Pt: 104 °C Melting Pt: 45 °C Flash Pt: 111 °C |
MDL Number:
MFCD00005445 CAS Number: 326-91-0 EINECS: 206-316-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...