Triethylenetetramine 60.0%

Supplier: TCI America
Danger

Synonyms: N,N'-Bis(2-aminoethyl)-1,2-ethanediamine, Trientine, DEH 24, EPH 925

T0429-500ML T0429-25ML
TCT0429-500MLEA 48.36 USD
TCT0429-500ML TCT0429-25ML
Triethylenetetramine 60.0%
Triethylenetetramine

CAS Number: 112-24-3
MDL Number: MFCD00008169
Molecular Formula: C6H18N4
Molecular Weight: 146.24
Purity/Analysis Method: >60.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 266
Flash Point (°C): 143
Specific Gravity (20/20): 0.98


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Monomers
Other Product Names / Synonyms: N,N'-Di(2-aminoethyl)ethylenediamine

Formula: C₆H₁₈N₄
MW: 146.24 g/mol
Boiling Pt: 266 °C
Density: 0.98 (20 °C)
Flash Pt: 143 °C
MDL Number: MFCD00008169
CAS Number: 112-24-3
EINECS: 203-950-6
UN: 2259
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbaca22a2357d563d08c97b56770c8899b5bcb87&vid=cbaca22a2357d563d08c97b56770c8899b5bcb87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >60.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR