Vinyltrichlorosilane ≥98.0%

Supplier: TCI America

Synonyms: Trichloro(vinyl)silane

T0407-25G T0407-100G
TCT0407-25GEA 23.51 USD
TCT0407-25G TCT0407-100G
Vinyltrichlorosilane ≥98.0%
Vinyltrichlorosilane

CAS Number: 75-94-5
MDL Number: MFCD00000482
Molecular Formula: C2H3Cl3Si
Molecular Weight: 161.48
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 92
Flash Point (°C): 18
Specific Gravity (20/20): 1.27

Formula: C₂H₃Cl₃Si
MW: 161.49 g/mol
Boiling Pt: 90
Melting Pt: -95
Density: 1.27 (20 °C)
Flash Pt: 18 °C
MDL Number: MFCD00000482
CAS Number: 75-94-5
EINECS: 200-917-8
UN: 1305
ADR: 3 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d24c0ef840c416e803ff40e53463bab12e7a1422&vid=d24c0ef840c416e803ff40e53463bab12e7a1422&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR