12-Tricosanone ≥95.0%

Supplier: TCI America

T0404-500G T0404-25G
TCT0404-500GEA 1011.6 USD
TCT0404-500G TCT0404-025G
12-Tricosanone ≥95.0%
12-Tricosanone

CAS Number: 540-09-0
MDL Number: MFCD00026562
Molecular Formula: C23H46O
Molecular Weight: 338.62
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 68


Other Product Names / Synonyms: Diundecyl Ketone, Laurone

Formula: C₂₃H₄₆O
MW: 338.62 g/mol
Melting Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00026562
CAS Number: 540-09-0
EINECS: 208-735-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdbbd92a500143a137658ca8ec03654003920444&vid=46ced39eaa0bc637dca749c83317d4c594615e10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR