2,4,6-Tribromophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
T0349-25G
T0349-500G
TCT0349-025GEA
21
USD
TCT0349-025G
TCT0349-500G
2,4,6-Tribromophenol ≥98.0% (by GC, titration analysis)
2,4,6-Tribromophenol
CAS Number: 118-79-6
MDL Number: MFCD00002150
Molecular Formula: C6H3Br3O
Molecular Weight: 330.80
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 244
Melting point (°C): 94
Formula:
C₆H₃Br₃O Boiling Pt: 244 °C Melting Pt: 94 °C Density: 2.55 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002150 CAS Number: 118-79-6 EINECS: 204-278-6 Merck Index: 12,09744 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...