2,4,6-Triallyloxy-1,3,5-triazine ≥98.0%

Supplier: TCI America

Synonyms: 2,4,6-Tris(allyloxy)-1,3,5-triazine, 2,4,6-Triallyloxy-s-triazine, 2,4,6-Tris(allyloxy)-s-triazine

T0333-500G T0333-25G
TCT0333-500GEA 137.49 USD
TCT0333-500G TCT0333-25G
2,4,6-Triallyloxy-1,3,5-triazine ≥98.0%
2,4,6-Triallyloxy-1,3,5-triazine

CAS Number: 101-37-1
MDL Number: MFCD00006049
Molecular Formula: C12H15N3O3
Molecular Weight: 249.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 162
Flash Point (°C): 168
Freezing point (°C): 27


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Cyanuric Acid Triallyl Ester

Formula: C₁₂H₁₅N₃O₃
Boiling Pt: 162 °C
Flash Pt: 168 °C
MDL Number: MFCD00006049
CAS Number: 101-37-1
EINECS: 202-936-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcad9eee80741db6edf109eaff092847310bbac8&vid=dcad9eee80741db6edf109eaff092847310bbac8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR