2-Toluoyl chloride ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
2-Methylbenzoyl chloride, o-Methylbenzoyl chloride, o-Toluoyl chloride, 2-Methylbenzene-1-carbonyl chloride
T0310-25G
T0310-100G
TCT0310-25GEA
27.4
USD
TCT0310-25G
TCT0310-100G
2-Toluoyl chloride ≥98.0%
2-Toluoyl chloride
CAS Number: 933-88-0
MDL Number: MFCD00000668
Molecular Formula: C8H7ClO
Molecular Weight: 154.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 213
Flash Point (°C): 101
Specific Gravity (20/20): 1.19
Other Product Names / Synonyms: 2-Methylbenzoyl Chloride
Formula:
C₈H₇ClO Boiling Pt: 213 °C Melting Pt: –18 °C Density: 1.19 (20 °C) Flash Pt: 101 °C |
MDL Number:
MFCD00000668 CAS Number: 933-88-0 EINECS: 213-273-8 UN: 3265 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...