p-Toluidine ≥99.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
4-Aminotoluene, 4-Methylaniline, 4-Toluidine
T0300-500G
T0300-25G
TCT0300-500GEA
84.42
USD
TCT0300-500G
TCT0300-25G
p-Toluidine ≥99.0% (by GC, titration analysis)
p-Toluidine
CAS Number: 106-49-0
MDL Number: MFCD00007906
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 201
Melting point (°C): 43
Flash Point (°C): 87
Storage Temperature: 0-10°C
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 201 °C Melting Pt: 43 °C Density: 1.046 g/cm³ (20 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007906 CAS Number: 106-49-0 EINECS: 203-403-1 UN: 3451 ADR: 6.1,II Merck Index: 12,09674 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...