m-Toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 3-Aminotoluene, 3-Toluidine, 3-Methylaniline

T0298-500ML T0298-25ML
TCT0298-500MLEA 147 USD
TCT0298-500ML TCT0298-25ML
m-Toluidine ≥98.0% (by GC, titration analysis)
m-Toluidine

CAS Number: 108-44-1
MDL Number: MFCD00007808
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 203
Flash Point (°C): 86
Specific Gravity (20/20): 0.99
Storage Temperature: 0-10°C

Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 203 °C
Melting Pt: -30 °C
Density: 0.99 (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00007808
CAS Number: 108-44-1
EINECS: 203-583-1
UN: 1708
ADR: 6.1,II
Merck Index: 13,09614
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6e753bf6f27b453867b1daa903484c4e47b96f8&vid=c6e753bf6f27b453867b1daa903484c4e47b96f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR