o-Toluic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Methylbenzoic acid

T0292-500G T0292-25G
TCT0292-500GEA 110 USD
TCT0292-500G TCT0292-025G
o-Toluic acid ≥98.0% (by titrimetric analysis)
o-Toluic acid

CAS Number: 118-90-1
MDL Number: MFCD00002477
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 259
Melting point (°C): 104
Flash Point (°C): 148


Other Product Names / Synonyms: 2-Methylbenzoic Acid

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 259 °C
Melting Pt: 104 °C
Density: 1.2555 g/cm³ (20 °C)
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00002477
CAS Number: 118-90-1
EINECS: 204-284-9
Merck Index: 12,09673
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44fba6356c8aa774509fd52ba72f7f9e1cd11814&vid=6756a4eceab66763be8464b58d8862ee92584bbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR