o-Toluenesulphonyl chloride ≥75.0% (contains ca. 23% isomer)

Supplier: TCI America
Danger

Synonyms: Toluene-o-sulphonyl chloride, Toluene-2-sulphonyl chloride, 2-Toluenesulphonyl chloride

T0284-500G T0284-25G
TCT0284-500GEA 226 USD
TCT0284-500G TCT0284-25G
o-Toluenesulphonyl chloride ≥75.0% (contains ca. 23% isomer)
o-Toluenesulphonyl chloride

CAS Number: 133-59-5
MDL Number: MFCD00024874
Molecular Formula: C7H7ClO2S
Molecular Weight: 190.64
Purity/Analysis Method: >75.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.34

Formula: C₇H₇ClO₂S
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 10.2 °C
Density: 1.34 (20 °C)
MDL Number: MFCD00024874
CAS Number: 133-59-5
EINECS: 205-113-0
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6942368e5055f1a41c266f5f87200ba5f733005e&vid=6942368e5055f1a41c266f5f87200ba5f733005e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >75.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR