Tiglic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(E)-2-Methylbut-2-enoic acid, trans-2,3-Dimethylacrylic acid
T0246-25G
T0246-100G
TCT0246-25GEA
44.78
USD
TCT0246-25G
TCT0246-100G
Tiglic acid ≥98.0% (by GC, titration analysis)
Tiglic acid
CAS Number: 80-59-1
MDL Number: MFCD00066864
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 82
Melting point (°C): 64
Flash Point (°C): 101
Storage Temperature: 0-10°C
Product Categories: Bioscience, Hemiterpenes
Other Product Names / Synonyms: (E)-2-Methyl-2-butenoic Acid, 2-Methylcrotonic Acid
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 82 °C Melting Pt: 64 °C Flash Pt: 101 °C |
MDL Number:
MFCD00066864 CAS Number: 80-59-1 EINECS: 201-295-0 UN: 3261 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...