2-Thiouracil ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Hydroxypyrimidine-2(1H)-thione

T0224-25G T0224-250G
TCT0224-025GEA 32.4 USD
TCT0224-025G TCT0224-250G
2-Thiouracil ≥98.0% (by HPLC, titration analysis)
2-Thiouracil

CAS Number: 141-90-2
MDL Number: MFCD00006039
Molecular Formula: C4H4N2OS
Molecular Weight: 128.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 331
Lambda max.: 271 nm (H2O)


Other Product Names / Synonyms: 2-Mercapto-4-hydroxypyrimidine, 2-Mercapto-4-pyrimidone

Formula: C₄H₄N₂OS
MW: 128.15 g/mol
Melting Pt: 331 °C
MDL Number: MFCD00006039
CAS Number: 141-90-2
EINECS: 205-508-8
Merck Index: 12,09504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b589923ef4ed909c7be2e1b6de392c09bb79fdeb&vid=b589923ef4ed909c7be2e1b6de392c09bb79fdeb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR