Thiophene ≥98.0%
Supplier: TCI America
|
Danger
|
CAS Number: 110-02-1
MDL Number: MFCD00005413
Molecular Formula: C4H4S
Molecular Weight: 84.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 84
Melting point (°C): -38
Flash Point (°C): -1
Specific Gravity (20/20): 1.07
Formula:
C₄H₄S Boiling Pt: 84 °C Melting Pt: –38 °C Density: 1.07 (20 °C) Flash Pt: –1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005413 CAS Number: 110-02-1 EINECS: 203-729-4 UN: 2414 ADR: 3,II Merck Index: 12,09490 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...