Thiosemicarbazide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: 3-Thiosemicarbazide

T0221-25G T0221-100G
TCT0221-25GEA 56.98 USD
TCT0221-25G TCT0221-100G
Thiosemicarbazide ≥98.0% (by titrimetric analysis)
Thiosemicarbazide

CAS Number: 79-19-6
MDL Number: MFCD00007620
Molecular Formula: CH5N3S
Molecular Weight: 91.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 184

Formula: CH₅N₃S
MW: 91.14 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00007620
CAS Number: 79-19-6
EINECS: 201-184-7
UN: 2811
ADR: 6.1,II
Merck Index: 12,09499
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d243ebb31b8f2ca25fce07cd0d6736c977787e6b&vid=d243ebb31b8f2ca25fce07cd0d6736c977787e6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR