(R)-(-)-Thiazolidine-4-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: H-Thz-OH, L(-)-Thiazolidine-4-carboxylic acid

T0219-500G T0219-25G
TCT0219-500GEA 412.11 USD
TCT0219-500G TCT0219-25G
(R)-(-)-Thiazolidine-4-carboxylic acid ≥98.0%
(R)-(-)-Thiazolidine-4-carboxylic acid

CAS Number: 34592-47-7
MDL Number: MFCD00005212
Molecular Formula: C4H7NO2S
Molecular Weight: 133.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Specific rotation [a]20/D: -200 deg (C=1, 1mol/L NaOH sol.)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Chiral Building Blocks, Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: (R)-(-)-Thiazolidine-4-carboxylic Acid

Formula: C₄H₇NO₂S
MDL Number: MFCD00005212
CAS Number: 34592-47-7
EINECS: 252-106-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=511f85689a1208048f18f186d095c866ac8a95b3&vid=511f85689a1208048f18f186d095c866ac8a95b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR