Thioacetic acid ≥95.0%
Supplier: TCI America
CAS Number: 507-09-5
MDL Number: MFCD00004853
Molecular Formula: C2H4OS
Molecular Weight: 76.11
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 93
Flash Point (°C): 5
Specific Gravity (20/20): 1.07
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Ethanethioic Acid
Formula:
C₂H₄OS Boiling Pt: 93 °C Melting Pt: –17 °C Density: 1.07 (20 °C) Flash Pt: 5 °C |
MDL Number:
MFCD00004853 CAS Number: 507-09-5 EINECS: 208-063-8 UN: 2436 ADR: 3,II Merck Index: 12,09454 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...